missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red TS, 0.1% (w/v) Alcoholic Solution, Ricca Chemical

Indicator for Acid-Base Titrations

Supplier:  Ricca Chemical Company 504516

 View more versions of this product

Catalog No. 504516


Only null left

Description

Description

Specifications

Chemical Identifiers

64-17-5 , 67-56-1 , 63451-28-5
100.0, 0.13
Indicator
pink (pH 4.2) - yellow (pH 6.2)
Red
Methyl Red, Test Solution
98.0, 0.12
0.1% (w/v)
Natural Poly Bottle
Alcoholic Solution
Liquid

Specifications

Methyl Red
64-17-5 , 67-56-1 , 63451-28-5
100.0, 0.13
Indicator
pink (pH 4.2) - yellow (pH 6.2)
Red
Liquid
Test Solution
98.0, 0.12
0.1% (w/v)
Natural Poly Bottle
Alcoholic Solution
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions