missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Solution R, Ricca Chemical

Catalog No. 50474
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50474 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50474 Supplier Ricca Chemical Company Supplier No. 50474
Only null left

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 67-56-1 , 493-52-7 , 7732-18-5
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) Mixture
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
PubChem CID 10303

Specifications

CAS Min % 55.0, 42.65, 0.05, 0.02
CAS Max % 58.0, 44.39, 0.06, 0.02
Grade Indicator
Packaging Amber Glass Bottle
Color Red
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Methyl Red Solution, Reagent