missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, sodium salt, ACS reagent

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00002426
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC414840100
View Documents Promotion Details
Thermo Scientific Chemicals
414840100
10 g Glass bottle N/A
Only null left
 
AC414840250
View Documents Promotion Details
Thermo Scientific Chemicals
414840250
25 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

845-10-3
291.286
GNTPCYMJCJNRQB-UHFFFAOYSA-M
4465632
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
C15H14N3NaO2
MFCD00002426
2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate

Specifications

845-10-3
C15H14N3NaO2
MFCD00002426
15, 6192
GNTPCYMJCJNRQB-UHFFFAOYSA-M
Authentic
291.286
291.27
From pH 4.2 (pink) to pH 6.2 (yellow)
Orange/Red to Yellow
Methyl Red, sodium salt, Pure
(Aqueous) Passes Test
(CH3)2NC6H4N=NC6H4CO2Na
16, 329
2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
4465632
ACS Reagent
13020
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-682-9

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only