Learn More
Thermo Scientific Chemicals Methyl Red, sodium salt, ACS reagent
CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
Supplier: Thermo Scientific Chemicals 414840250
| Quantity | 25 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 845-10-3 | |
| 291.286 | |
| GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
| 4465632 | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
| C15H14N3NaO2 | |
| MFCD00002426 | |
| 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR | |
| sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
Specifications
| Methyl Red, sodium salt | |
| 845-10-3 | |
| C15H14N3NaO2 | |
| MFCD00002426 | |
| 15, 6192 | |
| GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
| Authentic | |
| 291.286 | |
| 291.27 | |
| Glass bottle | |
| 13020 | |
| 25 g |
| Pure | |
| (Aqueous) Passes Test | |
| (CH3)2NC6H4N=NC6H4CO2Na | |
| 16, 329 | |
| 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] | |
| sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
| 4465632 | |
| ACS Reagent | |
| From pH 4.2 (pink) to pH 6.2 (yellow) | |
| Orange/Red to Yellow | |
| Liquid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 212-682-9
TSCA : TSCA