Learn More
Thermo Scientific Chemicals Methyl Red, sodium salt, pure, indicator
CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol
$77.34 - $1,019.25
Chemical Identifiers
| CAS | 845-10-3 |
|---|---|
| Molecular Formula | C15H14N3NaO2 |
| Molecular Weight (g/mol) | 291.286 |
| MDL Number | MFCD00002426 |
| InChI Key | GNTPCYMJCJNRQB-UHFFFAOYSA-M |
| Synonym | 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR |
| PubChem CID | 4465632 |
| IUPAC Name | sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC151410250
|
Thermo Scientific Chemicals
151410250 |
25 g | Glass bottle |
Each for $77.34
|
|
||||
|
AC151411000
|
Thermo Scientific Chemicals
151411000 |
100 g | Glass bottle |
Each for $211.79
|
|
||||
|
AC151415000
|
Thermo Scientific Chemicals
151415000 |
500 g | Glass bottle |
Each for $1,019.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 845-10-3 | |
| 291.286 | |
| GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
| 4465632 | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
| C15H14N3NaO2 | |
| MFCD00002426 | |
| 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR | |
| sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
Specifications
| 845-10-3 | |
| (CH3)2NC6H4N=NC6H4CO2Na | |
| 16, 329 | |
| 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] | |
| sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
| 4465632 | |
| Pure | |
| 13020 | |
| Flakes |
| C15H14N3NaO2 | |
| MFCD00002426 | |
| 15, 6192 | |
| GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
| Authentic | |
| 291.286 | |
| 291.27 | |
| From pH 4.2 (red) to pH 6.2 (yellow) | |
| Beige or White to Yellow | |
| Methyl Red, sodium salt, pure |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only