missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red-Methylene Blue TS, Alcoholic Mixed Indicator, Ricca Chemical
$156.90 - $156.90
Specifications
| Chemical Name or Material | Methyl Red-Methylene Blue |
|---|---|
| Name Note | Alcoholic Mixed Indicator, Test Solution |
| CAS | 64-17-5 , 67-56-1 , 493-52-7 , 7220-79-3 |
| CAS Min % | 98.0,0.06,0.03 |
| CAS Max % | 100.0,0.06,0.03 |