missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(+)-2-(4-Hydroxyphenoxy)propionate 98.0+%, TCI America™

Catalog No. H095625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H095625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H095625G Supplier TCI America Supplier No. H095625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96562-58-2
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00274087
InChI Key UUYSCNGPNOYZMC-SSDOTTSWSA-N
Synonym methyl r-+-2-4-hydroxyphenoxy propanoate, methyl r-+-2-4-hydroxyphenoxy propionate, r-methyl 2-4-hydroxyphenoxy propanoate, methyl r-2-4-hydroxyphenoxy propionate, r-+-2-4-hydroxyphenoxy propionic acid methyl ester, propanoic acid, 2-4-hydroxyphenoxy-, methyl ester, 2r, methyl 2r-2-4-hydroxyphenoxy propanoate, d-mhpp, aurora ka-6234
PubChem CID 2733752
IUPAC Name methyl (2R)-2-(4-hydroxyphenoxy)propanoate
SMILES COC(=O)[C@@H](C)OC1=CC=C(O)C=C1

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (R)-(+)-2-(4-Hydroxyphenoxy)propionate