accessibility menu, dialog, popup

UserName

H3COO(R)CH3OOH

CAS RN 96562-58-2

IUPAC Name: methyl (2R)-2-(4-hydroxyphenoxy)propanoate
Synonyms: methyl (2R)-2-(4-hydroxyphenoxy)propanoate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: UUYSCNGPNOYZMC-SSDOTTSWSA-N
SMILES: COC(=O)[C@@H](C)OC1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results