missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Purple Indicator Solution, Ricca Chemical
For acid-base titrations
$86.10 - $210.03
Specifications
| Chemical Name or Material | Methyl Purple Indicator Solution |
|---|---|
| CAS | 7732-18-5,845-10-3 |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Grade | Indicator |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
5040-4
|
Ricca Chemical Company
50404 |
120 mL | Amber Glass Bottle |
Each for $86.10
|
|
||||
|
5040-16
|
Ricca Chemical Company
504016 |
500 mL | Amber Glass Bottle |
Each for $140.33
|
|
||||
|
5040-32
|
Ricca Chemical Company
504032 |
1 L | Amber Glass Bottle |
Each for $210.03
|
|
||||
Specifications
| Methyl Purple Indicator Solution | |
| 98.0 | |
| Indicator | |
| Dark Green | |
| Liquid |
| 7732-18-5,845-10-3 | |
| 100.0 | |
| Amber Glass Bottle | |
| >5.4 |