Learn More
Methyl Purple Indicator Solution, Ricca Chemical
For acid-base titrations
Supplier:  Ricca Chemical Company 504016
 View more versions of this product
Catalog No. 5040-16
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Methyl Purple Indicator Solution
98.0
Indicator
pH 4.8 (purple) - 5.4 (green)
>5.4
Liquid
7732-18-5,845-10-3
100.0
Amber Glass Bottle
Dark Green
500 mL
Product Suggestions
Product Suggestions