missing translation for 'onlineSavingsMsg'
Learn More

Methyl Purple Indicator, pH 4.8 to 5.4 Purple to Green, Certified, LabChem™

Supplier:  LabChem LC170701

 View more versions of this product

Catalog No. LC170701

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Purple Indicator
84.9,15,0.06,0.04
C15H14N3O2Na
Passes Test
Certified
Sulfur compounds; Carbon monoxide; Carbon dioxide; Nitrogen oxides
500 mL
Liquid
pH 4.8 to 5.4 Purple to Green
67-63-0 , 3486-30-4 , 845-10-3
Soluble in water
291.28
Poly Bottle
Black/Green
33°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions