missing translation for 'onlineSavingsMsg'
Learn More

Methyl Phenyl Sulfone 97.0+%, TCI America™

Catalog No. p-7132144
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M040125G 25 g
M0401500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M040125G Supplier TCI America Supplier No. M040125G

Chemical Identifiers

CAS 3112-85-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00014741
InChI Key JCDWETOKTFWTHA-UHFFFAOYSA-N
Synonym methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone
PubChem CID 18369
IUPAC Name methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 88°C
Color White
Quantity 25 g
Formula Weight 156.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Phenyl Sulfone
RTECSNumber WR6800000
TSCA No