accessibility menu, dialog, popup

UserName

CH3SOO

CAS RN 3112-85-4

IUPAC Name: methanesulfonylbenzene
Synonyms: sulfone, phenyl, methyl-
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: JCDWETOKTFWTHA-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)