Learn More
Methyl phenyl sulfone, 99%
CAS: 3112-85-4 | C7H8O2S | 156.2 g/mol
$522.60 - $522.60
Chemical Identifiers
| CAS | 3112-85-4 |
|---|---|
| Molecular Formula | C7H8O2S |
| Molecular Weight (g/mol) | 156.2 |
| MDL Number | MFCD00014741 |
| InChI Key | JCDWETOKTFWTHA-UHFFFAOYSA-N |
| Synonym | methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone |
| PubChem CID | 18369 |
| IUPAC Name | methylsulfonylbenzene |
| SMILES | CS(=O)(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC353040250
|
Thermo Scientific Chemicals
353040250 |
25 g | Glass bottle |
Each for $522.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3112-85-4 | |
| 156.2 | |
| JCDWETOKTFWTHA-UHFFFAOYSA-N | |
| 18369 | |
| CS(=O)(=O)C1=CC=CC=C1 |
| C7H8O2S | |
| MFCD00014741 | |
| methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone | |
| methylsulfonylbenzene |
Specifications
| 3112-85-4 | |
| White | |
| 98.5% min. (GC) | |
| C7H8O2S | |
| MFCD00014741 | |
| methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone | |
| JCDWETOKTFWTHA-UHFFFAOYSA-N | |
| methylsulfonylbenzene | |
| 18369 | |
| 99% | |
| Methyl phenyl sulfone, 99% |
| 85°C to 88°C | |
| Authentic | |
| Glass bottle | |
| CH3SO2C6H5 | |
| 25 g | |
| Solubility in water: insoluble. Other solubilities: soluble in ethanol,benzene and chloroform,slightly soluble in carbon tetrachloride | |
| CS(=O)(=O)C1=CC=CC=C1 | |
| 156.2 | |
| 156.2 | |
| Crystals |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 221-476-8
RTECSNumber : WR6800000
RUO – Research Use Only