missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pentafluorophenyl Carbonate 98.0+%, TCI America™

Catalog No. M25941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25941G Supplier TCI America Supplier No. M25941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36919-03-6
Molecular Formula C8H3F5O3
Molecular Weight (g/mol) 242.10
MDL Number MFCD01075723
InChI Key HGYOVHMDBHQLOE-UHFFFAOYSA-N
Synonym methyl pentafluorophenyl carbonate, carbonic acid, methyl pentafluorophenyl ester, acmc-20akf4, pentafluorophenoxy methoxy ketone, methyl perfluorophenyl carbonate, carbonic acid methyl pentafluorophenyl ester
PubChem CID 14189360
IUPAC Name methyl 2,3,4,5,6-pentafluorophenyl carbonate
SMILES COC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 34°C
Color White
Boiling Point 84°C
Quantity 1 g
Formula Weight 242.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl Pentafluorophenyl Carbonate