accessibility menu, dialog, popup

UserName

H3COOOFFFFF

CAS RN 36919-03-6

IUPAC Name: methyl 2,3,4,5,6-pentafluorophenyl carbonate
Synonyms: methyl 2,3,4,5,6-pentafluorophenyl carbonate
Molecular Weight (g/mol): 242.1
Molecular Formula: C8H3F5O3
InChi Key: HGYOVHMDBHQLOE-UHFFFAOYSA-N
SMILES: COC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)