missing translation for 'onlineSavingsMsg'
Learn More

Methyl pentafluorophenyl carbonate, 97%, Thermo Scientific Chemicals

Catalog No. AAL1610706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1610706 5 g
AAL1610714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1610706 Supplier Thermo Scientific Chemicals Supplier No. L1610706

Methyl pentafluorophenyl carbonate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pentafluorophenyl carbonate

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 36919-03-6
Molecular Formula C8H3F5O3
Molecular Weight (g/mol) 242.10
MDL Number MFCD01075723
InChI Key HGYOVHMDBHQLOE-UHFFFAOYSA-N
Synonym methyl pentafluorophenyl carbonate, carbonic acid, methyl pentafluorophenyl ester, acmc-20akf4, pentafluorophenoxy methoxy ketone, methyl perfluorophenyl carbonate, carbonic acid methyl pentafluorophenyl ester
PubChem CID 14189360
SMILES COC(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 27°C to 31°C
Boiling Point 84°C (3 mmHg)
Quantity 5 g
UN Number UN2811
Beilstein 2289710
Solubility Information Insoluble in water.
IUPAC Name methyl (2,3,4,5,6-pentafluorophenyl) carbonate
Formula Weight 242.1
Percent Purity 97%
Chemical Name or Material Methyl pentafluorophenyl carbonate
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only