missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange Indicator, 0.2% (w/v), Ricca Chemical

$87.22 - $138.66

Specifications

Chemical Name or Material Methyl Orange Indicator
CAS 547-58-0,7732-18-5
CAS Min % 98.0,0.2
CAS Max % 100.0,0.2
Concentration 0.2% (w/v)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5020-4
View Documents
Ricca Chemical Company
50204
120 mL Natural Poly Bottle N/A
Only null left
 
5020-16
View Documents
Ricca Chemical Company
502016
500 mL Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Specifications

Methyl Orange Indicator
98.0,0.2
0.2% (w/v)
Indicator
Aqueous Solution
3 to 4
547-58-0,7732-18-5
100.0,0.2
0.2% W/V
Natural Poly Bottle
Orange
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'