missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator, 0.2% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 502016
Specifications
| Methyl Orange Indicator | |
| 98.0,0.2 | |
| 0.2% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 3 to 4 | |
| Liquid |
| 547-58-0,7732-18-5 | |
| 100.0,0.2 | |
| 0.2% W/V | |
| Natural Poly Bottle | |
| Orange | |
| 500 mL |