missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, 0.1% Aqueous, pH 3.2 to 4.4 Red to Yellow, LabChem™

Catalog No. LC170101
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC170101 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC170101 Supplier LabChem Supplier No. LC170101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified aqueous pH indicator for colorimetric analysis.

Chemical Identifiers

CAS 547-58-0 , 7732-18-5
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

CAS Max % 99.9, 0.1
Identification Passes Test
Packaging Poly Bottle
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides; Sulfur compounds
Color Orange
Solubility Information Soluble in water
pH 3.2 to 4.4
Quantity 500 mL
Physical Form Liquid
Formula Weight 327.34
Grade Certified
Chemical Name or Material Methyl Orange, 0.1% Aqueous
Show More Show Less
Recommended Storage Room Temperature