missing translation for 'onlineSavingsMsg'
Learn More

Methyl oleate, tech. C18 71-90%, C18:1 >65% of C18

CAS: 112-62-9 | C19H36O2 | 296.495 g/mol

Supplier:  Thermo Scientific Chemicals L02937AU

 View more versions of this product

Catalog No. AAL02937AU


Only null left

Description

Description

Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers, for duplicating ink, rubber and wax. It finds application as lubricant and lubricant additive.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Methyl oleate
0.876
163°C (325°F)
C18 71 to 90%, C18:1 >65% of C18
1.452
MFCD00009578
1727037
methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
QYDYPVFESGNLHU-KHPPLWFESA-N
methyl (Z)-octadec-9-enoate
5364509
296.5
112-62-9
168°C to 170°C (2 mmHg)
Weak, Characteristic
C19H36O2
CH3(CH2)7CH=CH(CH2)7CO2CH3
1000 mL
14,6828
Miscible with alcohol,ether and chloroform. Immiscible with water.
CCCCCCCCC=CCCCCCCCC(=O)OC
296.495
CHEBI:27542
Technical

Chemical Identifiers

112-62-9
296.495
QYDYPVFESGNLHU-KHPPLWFESA-N
5364509
methyl (Z)-octadec-9-enoate
C19H36O2
MFCD00009578
methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
CHEBI:27542
CCCCCCCCC=CCCCCCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-992-5

RTECSNumber : RK0895000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only