missing translation for 'onlineSavingsMsg'
Learn More

Methyl Oleate 60.0+%, TCI America™

Catalog No. p-7134159
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005525ML 25 mL
O0055500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O005525ML Supplier TCI America Supplier No. O005525ML
Only null left

Chemical Identifiers

CAS 112-62-9
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00009578
InChI Key QYDYPVFESGNLHU-KHPPLWFESA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 161°C
Quantity 25 mL
Formula Weight 296.50
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Oleate
EINECSNumber (2)-0798&(2)-0977
RTECSNumber RK0895000
TSCA Yes