missing translation for 'onlineSavingsMsg'
Learn More

Methyl Oleate 99.0+%, TCI America™

Catalog No. S03265ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03265ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03265ML Supplier TCI America Supplier No. S03265ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-62-9
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.495
MDL Number MFCD00009578
InChI Key QYDYPVFESGNLHU-KHPPLWFESA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -20°C
Color Colorless
Boiling Point 161°C
Quantity 5 mL
Formula Weight 296.50
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Oleate [Standard Material for GC]
EINECSNumber (2)-0798&(2)-0977
RTECSNumber RK0895000
TSCA Yes
Recommended Storage Refrigerator