missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl octanoate, 99%
CAS: 111-11-5 | C9H18O2 | 158.24 g/mol
Supplier: Thermo Scientific Chemicals 414621000
| Quantity | 100 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C9H18O2 | |
| MFCD00009551 | |
| methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
| CHEBI:87432 | |
| CCCCCCCC(=O)OC |
Specifications
| Methyl octanoate | |
| 1mg KOH/g max. | |
| Colorless | |
| 79.0°C (12.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4165 to 1.4185 | |
| 100 g | |
| 0.878 | |
| Solubility in water: insoluble | |
| CCCCCCCC(=O)OC | |
| 158.24 | |
| CHEBI:87432 | |
| 99% |
| 99% | |
| 111-11-5 | |
| 0.8780g/mL | |
| 73°C | |
| 98.5% min. (GC) | |
| C9H18O2 | |
| CH3(CH2)6COOCH3 | |
| MFCD00009551 | |
| methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
| JGHZJRVDZXSNKQ-UHFFFAOYSA-N | |
| methyl octanoate | |
| 8091 | |
| 158.24 | |
| Liquid |
Safety and Handling
EINECSNumber : 203-835-0
RTECSNumber : RH0778000
TSCA : TSCA