missing translation for 'onlineSavingsMsg'
Learn More

Methyl nervonate, 99%, analytical standard for GC

CAS: 2733-88-2 | C25H48O2 | 380.65 g/mol

Chemical Identifiers

CAS 2733-88-2
Molecular Formula C25H48O2
Molecular Weight (g/mol) 380.65
InChI Key AINIZSBLAFHZCP-KHPPLWFESA-N
Synonym nervonic acid methyl ester, methyl nervonate, methyl z-tetracos-15-enoate, methyl cis-15-tetracosenoate, selacholeic acid methyl ester, 15-tetracosenoic acid, methyl ester, z, methyl nervonate c24:1, z-methyl tetracos-15-enoate, methyl 15z tetracos-15-enoate, methyl 15z-15-tetracosenoate #
PubChem CID 5364841
IUPAC Name methyl (Z)-tetracos-15-enoate
SMILES CCCCCCCCC=CCCCCCCCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC460371000
View Documents Promotion Details
N/A 100 mg N/A
Only null left
 
AC460375000
View Documents Promotion Details
N/A 500 mg N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2733-88-2
380.65
nervonic acid methyl ester, methyl nervonate, methyl z-tetracos-15-enoate, methyl cis-15-tetracosenoate, selacholeic acid methyl ester, 15-tetracosenoic acid, methyl ester, z, methyl nervonate c24:1, z-methyl tetracos-15-enoate, methyl 15z tetracos-15-enoate, methyl 15z-15-tetracosenoate #
methyl (Z)-tetracos-15-enoate
C25H48O2
AINIZSBLAFHZCP-KHPPLWFESA-N
5364841
CCCCCCCCC=CCCCCCCCCCCCCCC(=O)OC

Specifications

2733-88-2
98.5% min. (GC)
CH3(CH2)7CHCH(CH2)13COOCH3
AINIZSBLAFHZCP-KHPPLWFESA-N
methyl (Z)-tetracos-15-enoate
5364841
99%
Authentic
C25H48O2
nervonic acid methyl ester, methyl nervonate, methyl z-tetracos-15-enoate, methyl cis-15-tetracosenoate, selacholeic acid methyl ester, 15-tetracosenoic acid, methyl ester, z, methyl nervonate c24:1, z-methyl tetracos-15-enoate, methyl 15z tetracos-15-enoate, methyl 15z-15-tetracosenoate #
CCCCCCCCC=CCCCCCCCCCCCCCC(=O)OC
380.65
380.65
Methyl nervonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-352-

SDS
Documents

Documents

RUO – Research Use Only