missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Myristate 98.0+%, TCI America™
$37.97 - $104.43
Chemical Identifiers
| CAS | 124-10-7 |
|---|---|
| Molecular Formula | C15H30O2 |
| Molecular Weight (g/mol) | 242.40 |
| MDL Number | MFCD00008983 |
| InChI Key | ZAZKJZBWRNNLDS-UHFFFAOYSA-N |
| Synonym | methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci |
| PubChem CID | 31284 |
| IUPAC Name | methyl tetradecanoate |
| SMILES | CCCCCCCCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
M048225ML
|
N/A | 25 mL |
Each for $37.97
|
|
|||||
|
M0482500ML
|
N/A | 500 mL |
Each for $104.43
|
|
|||||
Chemical Identifiers
| C15H30O2 | |
| MFCD00008983 | |
| methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci | |
| methyl tetradecanoate |
Specifications
| 124-10-7 | |
| 173°C | |
| MFCD00008983 | |
| ZAZKJZBWRNNLDS-UHFFFAOYSA-N | |
| methyl tetradecanoate | |
| 31284 | |
| ≥98.0% (GC) | |
| Methyl Myristate |
| Yellow | |
| C15H30O2 | |
| methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci | |
| CCCCCCCCCCCCCC(=O)OC | |
| 242.40 | |
| 242.40 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes