missing translation for 'onlineSavingsMsg'
Learn More

Methyl Myristate 98.0+%, TCI America™

Catalog No. M048225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M048225ML 25 mL
M0482500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M048225ML Supplier TCI America Supplier No. M048225ML

Chemical Identifiers

CAS 124-10-7
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.40
MDL Number MFCD00008983
InChI Key ZAZKJZBWRNNLDS-UHFFFAOYSA-N
Synonym methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
PubChem CID 31284
IUPAC Name methyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 mL
Formula Weight 242.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Myristate
EINECSNumber (2)-0798
TSCA Yes