missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl myristate, ≥99.5% (GC), MilliporeSigma™ Supelco™
Methyl myristate is generally used with methyl laurate as lubricant for textiles, metalworking and agricultural applications.
$54.03 - $279.24
Specifications
| CAS | 124-10-7 |
|---|---|
| Density | 0.855 g/mL (at 25°C) |
| Flash Point | 112°C |
| Shipping Condition | Ambient |
| Physical Form | Neat |
Specifications
| 124-10-7 | |
| 112°C | |
| Neat | |
| MFCD00008983 | |
| 323°C | |
| Analytical Standard | |
| ≥99.5% (GC) |
| 0.855 g/mL (at 25°C) | |
| Ambient | |
| CH3(CH2)12 COOCH3 | |
| 18°C | |
| Methyl tetradecanoate; Myristic acid methyl ester | |
| 242.40 | |
| Analytical Standard |