missing translation for 'onlineSavingsMsg'
Learn More

Methyl myristate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012828
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012828 1 mL
111012829 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012828 Supplier Merck Emd Millipore Supplier No. 701291ML
Only null left

Methyl myristate is generally used with methyl laurate as lubricant for textiles, metalworking and agricultural applications.

Specifications

CAS 124-10-7
Density 0.855 g/mL (at 25°C)
Flash Point 112°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)12 COOCH3
MDL Number MFCD00008983
Melting Point 18°C
Boiling Point 323°C
Synonym Methyl tetradecanoate; Myristic acid methyl ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 242.40
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less