missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Lignocerate 98.0+%, TCI America™
$233.00 - $622.59
Chemical Identifiers
| CAS | 2442-49-1 |
|---|---|
| Molecular Formula | C25H50O2 |
| Molecular Weight (g/mol) | 382.673 |
| MDL Number | MFCD00009351 |
| InChI Key | XUDJZDNUVZHSKZ-UHFFFAOYSA-N |
| Synonym | methyl lignocerate, lignoceric acid methyl ester, tetracosanoic acid, methyl ester, tetracosanoic acid methyl ester, methyl tetracosanoic acid, unii-hy5564b8fx, lignoceric methyl ester, tetracosanoic acid methyl, acmc-209gc6, tetracosanoic acid,methyl ester |
| PubChem CID | 75546 |
| IUPAC Name | methyl tetracosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC |
Chemical Identifiers
| 2442-49-1 | |
| 382.673 | |
| XUDJZDNUVZHSKZ-UHFFFAOYSA-N | |
| 75546 | |
| CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC |
| C25H50O2 | |
| MFCD00009351 | |
| methyl lignocerate, lignoceric acid methyl ester, tetracosanoic acid, methyl ester, tetracosanoic acid methyl ester, methyl tetracosanoic acid, unii-hy5564b8fx, lignoceric methyl ester, tetracosanoic acid methyl, acmc-209gc6, tetracosanoic acid,methyl ester | |
| methyl tetracosanoate |
Specifications
| 2442-49-1 | |
| White | |
| C25H50O2 | |
| 1 g | |
| XUDJZDNUVZHSKZ-UHFFFAOYSA-N | |
| methyl tetracosanoate | |
| 75546 | |
| ≥98.0% (GC) | |
| Methyl Lignocerate |
| 60°C | |
| 232°C | |
| MFCD00009351 | |
| methyl lignocerate, lignoceric acid methyl ester, tetracosanoic acid, methyl ester, tetracosanoic acid methyl ester, methyl tetracosanoic acid, unii-hy5564b8fx, lignoceric methyl ester, tetracosanoic acid methyl, acmc-209gc6, tetracosanoic acid,methyl ester | |
| CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC | |
| 382.673 | |
| 382.67 | |
| Crystalline Powder |
Safety and Handling
TSCA : No