missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Lignocerate 98.0+%, TCI America™
Supplier: TCI America L01125G
Specifications
| Methyl Lignocerate | |
| 60°C | |
| 232°C | |
| MFCD00009351 | |
| methyl lignocerate, lignoceric acid methyl ester, tetracosanoic acid, methyl ester, tetracosanoic acid methyl ester, methyl tetracosanoic acid, unii-hy5564b8fx, lignoceric methyl ester, tetracosanoic acid methyl, acmc-209gc6, tetracosanoic acid,methyl ester | |
| CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC | |
| 382.673 | |
| 382.67 | |
| Crystalline Powder |
| 2442-49-1 | |
| White | |
| C25H50O2 | |
| 5 g | |
| XUDJZDNUVZHSKZ-UHFFFAOYSA-N | |
| methyl tetracosanoate | |
| 75546 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2442-49-1 | |
| 382.673 | |
| XUDJZDNUVZHSKZ-UHFFFAOYSA-N | |
| 75546 | |
| CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC |
| C25H50O2 | |
| MFCD00009351 | |
| methyl lignocerate, lignoceric acid methyl ester, tetracosanoic acid, methyl ester, tetracosanoic acid methyl ester, methyl tetracosanoic acid, unii-hy5564b8fx, lignoceric methyl ester, tetracosanoic acid methyl, acmc-209gc6, tetracosanoic acid,methyl ester | |
| methyl tetracosanoate |
Safety and Handling
TSCA : No