missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Jasmonate (mixture of isomers) 90.0+%, TCI America™
Supplier: TCI America M106825G
Specifications
| Methyl Jasmonate (mixture of isomers) | |
| Yellow | |
| C13H20O3 | |
| 25 g | |
| GEWDNTWNSAZUDX-UHFFFAOYNA-N | |
| methyl 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetate | |
| 5319693 | |
| ≥90.0% (GC) |
| 1101843-02-0 | |
| 96°C | |
| MFCD00151382 | |
| Jasmonic Acid Methyl Ester, Methyl 3-Oxo-2-(2-pentenyl)cyclopentaneacetate, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, 2-(2-Pentenyl)-3-methoxycarbonylmethylcyclopentanone | |
| CCC=CCC1C(CC(=O)OC)CCC1=O | |
| 224.30 | |
| 224.30 | |
| Liquid |
Chemical Identifiers
| 1101843-02-0 | |
| 224.30 | |
| GEWDNTWNSAZUDX-UHFFFAOYNA-N | |
| 5319693 | |
| CCC=CCC1C(CC(=O)OC)CCC1=O |
| C13H20O3 | |
| MFCD00151382 | |
| Jasmonic Acid Methyl Ester, Methyl 3-Oxo-2-(2-pentenyl)cyclopentaneacetate, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, 2-(2-Pentenyl)-3-methoxycarbonylmethylcyclopentanone | |
| methyl 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetate |
Safety and Handling
EINECSNumber : (3)-3839
TSCA : Yes