CAS RN 1101843-02-0
IUPAC Name:
methyl 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetate
Synonyms:
methyl 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetate
Molecular Weight (g/mol):
224.3
Molecular Formula:
C13H20O3
InChi Key:
GEWDNTWNSAZUDX-UHFFFAOYNA-N
SMILES:
CCC=CCC1C(CC(=O)OC)CCC1=O
1
–
1
of
1
results
1
Methyl Jasmonate (mixture of isomers) 90.0+%, TCI America™
CAS: 1101843-02-0 Molecular Formula: C13H20O3 Molecular Weight (g/mol): 224.30 MDL Number: MFCD00151382 InChI Key: GEWDNTWNSAZUDX-UHFFFAOYNA-N Synonym: Jasmonic Acid Methyl Ester, Methyl 3-Oxo-2-(2-pentenyl)cyclopentaneacetate, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, 2-(2-Pentenyl)-3-methoxycarbonylmethylcyclopentanone PubChem CID: 5319693 IUPAC Name: methyl 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetate SMILES: CCC=CCC1C(CC(=O)OC)CCC1=O