missing translation for 'onlineSavingsMsg'
Learn More

Methyl Jasmonate (mixture of isomers) 90.0+%, TCI America™

Catalog No. p-7132522
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10685G 5 g
M106825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10685G Supplier TCI America Supplier No. M10685G

Chemical Identifiers

CAS 1101843-02-0
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.30
MDL Number MFCD00151382
InChI Key GEWDNTWNSAZUDX-UHFFFAOYNA-N
Synonym Jasmonic Acid Methyl Ester, Methyl 3-Oxo-2-(2-pentenyl)cyclopentaneacetate, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, 2-(2-Pentenyl)-3-methoxycarbonylmethylcyclopentanone
PubChem CID 5319693
IUPAC Name methyl 2-[3-oxo-2-(pent-2-en-1-yl)cyclopentyl]acetate
SMILES CCC=CCC1C(CC(=O)OC)CCC1=O

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 224.30
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Jasmonate (mixture of isomers)
EINECSNumber (3)-3839
TSCA Yes