missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Hexanoate 99.5+%, TCI America™
Supplier: TCI America S03045ML
Specifications
| Methyl Hexanoate [Standard Material for GC] | |
| Colorless | |
| C7H14O2 | |
| 5 mL | |
| methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate | |
| CCCCCC(=O)OC | |
| 130.19 | |
| CHEBI:77322 | |
| ≥99.5% (GC) |
| 106-70-7 | |
| 150°C | |
| MFCD00009510 | |
| 3272 | |
| NUKZAGXMHTUAFE-UHFFFAOYSA-N | |
| methyl hexanoate | |
| 7824 | |
| 130.19 | |
| Liquid |
Chemical Identifiers
| 106-70-7 | |
| 130.19 | |
| NUKZAGXMHTUAFE-UHFFFAOYSA-N | |
| 7824 | |
| methyl hexanoate |
| C7H14O2 | |
| MFCD00009510 | |
| methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate | |
| CHEBI:77322 | |
| CCCCCC(=O)OC |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : MO8401400
TSCA : Yes