missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl heptadecanoate, ≥99% (GC), MilliporeSigma™ Supelco™
$184.00 - $540.53
Specifications
| CAS | 1731-92-6 |
|---|---|
| Flash Point | >110°C |
| Shipping Condition | Ambient |
| Physical Form | Neat |
| Linear Formula | CH3(CH2)15 COOCH3 |
Specifications
| 1731-92-6 | |
| Ambient | |
| CH3(CH2)15 COOCH3 | |
| 29.8°C to 30.3°C | |
| Heptadecanoic acid methyl ester; Methyl margarate | |
| 284.48 | |
| Analytical Standard |
| >110°C | |
| Neat | |
| MFCD00009001 | |
| 152°C to 153°C (0.05 mmHg) | |
| Analytical Standard | |
| ≥99% (GC) |