missing translation for 'onlineSavingsMsg'
Learn More

Methyl heptadecanoate, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013344
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013344 1 g
111013345 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013344 Supplier Merck Emd Millipore Supplier No. 516331G
Only null left

Specifications

CAS 1731-92-6
Flash Point >110°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)15 COOCH3
MDL Number MFCD00009001
Melting Point 29.8°C to 30.3°C
Boiling Point 152°C to 153°C (0.05 mmHg)
Synonym Heptadecanoic acid methyl ester; Methyl margarate
Type Analytical Standard
Quantity 1 g
Formula Weight 284.48
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less