missing translation for 'onlineSavingsMsg'
Learn More

Methyl(diphenyl)phosphine Oxide 98.0+%, TCI America™

Catalog No. d244510g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D244510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D244510G Supplier TCI America Supplier No. D244510G

Chemical Identifiers

CAS 2129-89-7
Molecular Formula C13H13OP
Molecular Weight (g/mol) 216.22
MDL Number MFCD00002081
InChI Key PEGCITODQASXKH-UHFFFAOYSA-N
Synonym methyldiphenylphosphine oxide, methyl diphenyl phosphine oxide, diphenylmethylphosphine oxide, phosphine oxide, methyldiphenyl, phosphine oxide, diphenylmethyl, methyl-phenylphosphoryl benzene, methyl phenyl phosphoryl benzene, methyl phenyl phosphoroso benzene, methyldiphenylphosphineoxide
PubChem CID 75041
IUPAC Name [methyl(phenyl)phosphoroso]benzene
SMILES CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 113°C
Color White-Yellow
Boiling Point 180°C
Quantity 10 g
Formula Weight 216.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl(diphenyl)phosphine Oxide
RTECSNumber SZ0199480
TSCA No