accessibility menu, dialog, popup

UserName

CH3PO

CAS RN 2129-89-7

IUPAC Name: [methyl(phenyl)phosphoroso]benzene
Synonyms: [methyl(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 216.22
Molecular Formula: C13H13OP
InChi Key: PEGCITODQASXKH-UHFFFAOYSA-N
SMILES: CP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results