missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Decanoate 98.0+%, TCI America™
$46.60 - $134.21
Chemical Identifiers
| CAS | 110-42-9 |
|---|---|
| Molecular Formula | C11H22O2 |
| Molecular Weight (g/mol) | 186.30 |
| MDL Number | MFCD00009580 |
| InChI Key | YRHYCMZPEVDGFQ-UHFFFAOYSA-N |
| Synonym | methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate |
| PubChem CID | 8050 |
| IUPAC Name | methyl decanoate |
| SMILES | CCCCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D002325ML
|
TCI America
D002325ML |
25 mL |
Each for $46.60
|
|
|||||
|
D0023500ML
|
TCI America
D0023500ML |
500 mL |
Each for $134.21
|
|
|||||
Chemical Identifiers
| 110-42-9 | |
| 186.30 | |
| YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
| 8050 | |
| CCCCCCCCCC(=O)OC |
| C11H22O2 | |
| MFCD00009580 | |
| methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
| methyl decanoate |
Specifications
| 110-42-9 | |
| Colorless | |
| C11H22O2 | |
| 25 mL | |
| YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
| methyl decanoate | |
| 8050 | |
| ≥98.0% (GC) | |
| Methyl Decanoate |
| -18°C | |
| 229°C | |
| MFCD00009580 | |
| methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
| CCCCCCCCCC(=O)OC | |
| 186.30 | |
| 186.30 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0782
RTECSNumber : HD9830000
TSCA : Yes