Learn More
Methyl decanoate, 98+%
CAS: 110-42-9 | C11H22O2 | 186.30 g/mol
Supplier: Thermo Scientific Chemicals A1565822
Description
It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Methyl decanoate | |
| -18°C | |
| 223°C to 225°C | |
| C11H22O2 | |
| MFCD00009580 | |
| 1759170 | |
| Soluble in alcohol. Insoluble in water. | |
| CCCCCCCCCC(=O)OC | |
| 186.30 | |
| 186.3 |
| 110-42-9 | |
| 0.877 | |
| 94°C (201°F) | |
| 1.425 | |
| 100 g | |
| methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
| YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
| methyl decanoate | |
| 8050 | |
| 99% |
Chemical Identifiers
| 110-42-9 | |
| 186.30 | |
| YRHYCMZPEVDGFQ-UHFFFAOYSA-N | |
| 8050 | |
| CCCCCCCCCC(=O)OC |
| C11H22O2 | |
| MFCD00009580 | |
| methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate | |
| methyl decanoate |
Safety and Handling
P210-P233-P240-P241-P242-P243-P280-P374-P501c
H227
EINECSNumber : 203-766-6
RTECSNumber : HD9830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only