missing translation for 'onlineSavingsMsg'
Learn More

Methyl decanoate, 98+%

Catalog No. AAA1565818
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1565818 50 g
AAA1565822 100 g
AAA1565830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1565818 Supplier Thermo Scientific Chemicals Supplier No. A1565818
Only null left

CAS: 110-42-9 | C11H22O2 | 186.30 g/mol

It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 110-42-9
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009580
InChI Key YRHYCMZPEVDGFQ-UHFFFAOYSA-N
Synonym methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate
PubChem CID 8050
IUPAC Name methyl decanoate
SMILES CCCCCCCCCC(=O)OC

Specifications

Melting Point -18°C
Density 0.877
Boiling Point 223°C to 225°C
Flash Point 94°C (201°F)
Refractive Index 1.425
Quantity 50 g
Beilstein 1759170
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 186.3
Percent Purity 99%
Chemical Name or Material Methyl decanoate
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P374-P501c
EINECSNumber 203-766-6
RTECSNumber HD9830000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only