missing translation for 'onlineSavingsMsg'
Learn More

Methyl cis-9-Hexadecenoate 98.0+%, TCI America™

Catalog No. P1958100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1958100MG 100 mg
P19581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1958100MG Supplier TCI America Supplier No. P1958100MG

Chemical Identifiers

CAS 1120-25-8
Molecular Formula C17H32O2
Molecular Weight (g/mol) 268.44
MDL Number MFCD00042911
InChI Key IZFGRAGOVZCUFB-HJWRWDBZSA-N
Synonym methyl palmitoleate, z-methyl hexadec-9-enoate, methyl z-hexadec-9-enoate, methyl palmitoleinate, unii-35695qdb9f, methyl 9z-hexadec-9-enoate, 9-hexadecenoic acid, methyl ester, z, methyl cis-9-hexadecenoate, methyl z-9-hexadecenoate, methyl cis-hexadec-9-enoate
PubChem CID 643801
ChEBI CHEBI:84156
IUPAC Name methyl (9Z)-hexadec-9-enoate
SMILES CCCCCC\C=C/CCCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 126°C
Quantity 100 mg
Formula Weight 268.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl cis-9-Hexadecenoate
EINECSNumber (2)-0798
TSCA No
Recommended Storage Freezer