missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl cis-13-Docosenoate 90.0+%, TCI America™
$209.84 - $1803.89
Chemical Identifiers
| CAS | 1120-34-9 |
|---|---|
| Molecular Formula | C23H44O2 |
| Molecular Weight (g/mol) | 352.603 |
| MDL Number | MFCD00027343 |
| InChI Key | ZYNDJIBBPLNPOW-KHPPLWFESA-N |
| Synonym | methyl erucate, erucic acid methyl ester, methyl cis-13-docosenoate, z-methyl docos-13-enoate, methyl z-docos-13-enoate, methyl z-13 docosenoate, unii-dt8r52277s, 13-docosenoic acid, methyl ester, z, methyl 13z-docos-13-enoate, 13-docosenoic acid, methyl ester, 13z |
| PubChem CID | 5364423 |
| IUPAC Name | methyl (Z)-docos-13-enoate |
| SMILES | CCCCCCCCC=CCCCCCCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D101725ML
|
TCI America
D101725ML |
25 mL |
Each for $209.84
|
|
|||||
|
D1017500ML
|
TCI America
D1017500ML |
500 mL |
Each for $1,803.89
|
|
|||||
Chemical Identifiers
| 1120-34-9 | |
| 352.603 | |
| ZYNDJIBBPLNPOW-KHPPLWFESA-N | |
| 5364423 | |
| CCCCCCCCC=CCCCCCCCCCCCC(=O)OC |
| C23H44O2 | |
| MFCD00027343 | |
| methyl erucate, erucic acid methyl ester, methyl cis-13-docosenoate, z-methyl docos-13-enoate, methyl z-docos-13-enoate, methyl z-13 docosenoate, unii-dt8r52277s, 13-docosenoic acid, methyl ester, z, methyl 13z-docos-13-enoate, 13-docosenoic acid, methyl ester, 13z | |
| methyl (Z)-docos-13-enoate |
Specifications
| 1120-34-9 | |
| 222°C | |
| MFCD00027343 | |
| 3272 | |
| ZYNDJIBBPLNPOW-KHPPLWFESA-N | |
| methyl (Z)-docos-13-enoate | |
| 5364423 | |
| ≥90.0% (GC) | |
| Methyl cis-13-Docosenoate |
| Yellow | |
| C23H44O2 | |
| 25 mL | |
| methyl erucate, erucic acid methyl ester, methyl cis-13-docosenoate, z-methyl docos-13-enoate, methyl z-docos-13-enoate, methyl z-13 docosenoate, unii-dt8r52277s, 13-docosenoic acid, methyl ester, z, methyl 13z-docos-13-enoate, 13-docosenoic acid, methyl ester, 13z | |
| CCCCCCCCC=CCCCCCCCCCCCC(=O)OC | |
| 352.603 | |
| 352.60 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes