missing translation for 'onlineSavingsMsg'
Learn More

Methyl cis-11-eicosenoate, 99%

Catalog No. p-4089640
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428530010 1 g Glass bottle
AC428530050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428530010 Supplier Thermo Scientific Chemicals Supplier No. 428530010

CAS: 2390-09-2 | C21H40O2 | 324.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2390-09-2
Molecular Formula C21H40O2
Molecular Weight (g/mol) 324.55
MDL Number MFCD00010458
InChI Key RBKMRGOHCLRTLZ-ZHACJKMWSA-N
Synonym cis-11-eicosenoic acid methyl ester, cis-methyl 11-eicosenoate, 11-eicosenoic acid, methyl ester, z, methyl cis-11-eicosenoate, methyl 11z-icos-11-enoate, methyl z-icos-11-enoate, z-methyl icos-11-enoate, cis-11-eicosenoic acid, methyl ester, 11z-11-icosenoic acid methyl ester
PubChem CID 5463047
IUPAC Name methyl (Z)-icos-11-enoate
SMILES CCCCCCCCC=CCCCCCCCCCC(=O)OC

Specifications

Melting Point -15.0°C
Flash Point >113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH=CH(CH2)9CO2CH3
Quantity 1 g
Formula Weight 324.55
Percent Purity 99%
Chemical Name or Material Methyl cis-11-eicosenoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 219-226-8

RUO – Research Use Only