missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl cinnamate, predominantly trans, 99%
CAS: 103-26-4 | C10H10O2 | 162.188 g/mol
$60.38 - $925.01
Chemical Identifiers
| CAS | 103-26-4 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.188 |
| MDL Number | MFCD00008458 |
| InChI Key | CCRCUPLGCSFEDV-BQYQJAHWSA-N |
| Synonym | methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate |
| PubChem CID | 637520 |
| ChEBI | CHEBI:6857 |
| IUPAC Name | methyl (E)-3-phenylprop-2-enoate |
| SMILES | COC(=O)C=CC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1597522
|
Thermo Scientific Chemicals
A1597522 |
100 g |
N/A
|
|
|||||
|
AAA1597536
|
Thermo Scientific Chemicals
A1597536 |
500 g |
N/A
|
|
|||||
|
AAA159750E
|
Thermo Scientific Chemicals
A159750E |
2500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsMethyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 103-26-4 | |
| 162.188 | |
| CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
| 637520 | |
| methyl (E)-3-phenylprop-2-enoate |
| C10H10O2 | |
| MFCD00008458 | |
| methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
| CHEBI:6857 | |
| COC(=O)C=CC1=CC=CC=C1 |
Specifications
| 103-26-4 | |
| 1.042 | |
| 123°C (253°F) | |
| C10H10O2 | |
| MFCD00008458 | |
| 386468 | |
| methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate | |
| CCRCUPLGCSFEDV-BQYQJAHWSA-N | |
| methyl (E)-3-phenylprop-2-enoate | |
| 637520 | |
| 162.19 | |
| Methyl Cinnamate, predominantly trans |
| 34°C to 37°C | |
| 262°C to 264°C | |
| 99% | |
| 1.577 | |
| 100 g | |
| 14,2299 | |
| Insoluble in water. | |
| COC(=O)C=CC1=CC=CC=C1 | |
| 162.188 | |
| CHEBI:6857 | |
| 99% |
Safety and Handling
EINECSNumber : 203-093-8
RTECSNumber : GE0190000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only