missing translation for 'onlineSavingsMsg'
Learn More

Methyl cinnamate, predominantly trans, 99%

CAS: 103-26-4 | C10H10O2 | 162.188 g/mol

Supplier:  Thermo Scientific Chemicals A159750E

 View more versions of this product

Catalog No. AAA159750E


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl cinnamate
103-26-4
1.042
123°C (253°F)
C10H10O2
MFCD00008458
386468
methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
CCRCUPLGCSFEDV-BQYQJAHWSA-N
methyl (E)-3-phenylprop-2-enoate
637520
162.19
predominantly trans
34°C to 37°C
262°C to 264°C
99%
1.577
2500 g
14,2299
Insoluble in water.
COC(=O)C=CC1=CC=CC=C1
162.188
CHEBI:6857
99%

Chemical Identifiers

103-26-4
162.188
CCRCUPLGCSFEDV-BQYQJAHWSA-N
637520
methyl (E)-3-phenylprop-2-enoate
C10H10O2
MFCD00008458
methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
CHEBI:6857
COC(=O)C=CC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-093-8

RTECSNumber : GE0190000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only