Learn More
Thermo Scientific Chemicals Methyl cellulose, viscosity 8000 cPs
CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
$120.48 - $525.77
Chemical Identifiers
| CAS | 9004-67-5 |
|---|---|
| Molecular Formula | C20H38O11 |
| Molecular Weight (g/mol) | 454.513 |
| MDL Number | MFCD00081763 |
| InChI Key | YLGXILFCIXHCMC-JHGZEJCSSA-N |
| PubChem CID | 51063134 |
| IUPAC Name | (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| SMILES | COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Description
Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. It is also used as binders, lubricants, steric stabilizer, water retention aid and sizing in the production of papers and textiles as it protects the fibers from absorbing water or oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9004-67-5 | |
| 454.513 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| C20H38O11 | |
| MFCD00081763 | |
| 51063134 | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Specifications
| 9004-67-5 | |
| C20H38O11 | |
| 14,6040 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane | |
| 51063134 | |
| Methyl cellulose, Viscosity: 8000cPs |
| Odorless | |
| MFCD00081763 | |
| Soluble in glacial acetic acid and cold water. Insoluble in hot water,ethanol,ether and chloroform. | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC | |
| 454.513 | |
| Powder |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only