missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl cellulose, viscosity 8000 cPs

Catalog No. AA4348322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4348322 100 g
AA4348336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4348322 Supplier Thermo Scientific Chemicals Supplier No. 04348322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol

Methyl cellulose is used as a thickener and emulsifier in various food and cosmetic products, and also as a treatment of constipation. It is also used as binders, lubricants, steric stabilizer, water retention aid and sizing in the production of papers and textiles as it protects the fibers from absorbing water or oil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
MDL Number MFCD00081763
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Odor Odorless
Quantity 100 g
Merck Index 14,6040
Solubility Information Soluble in glacial acetic acid and cold water. Insoluble in hot water,ethanol,ether and chloroform.
Physical Form Powder
Chemical Name or Material Methyl cellulose, Viscosity: 8000cPs
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only