Learn More
Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cPs
CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
$73.99 - $950.56
Chemical Identifiers
| CAS | 9004-67-5 |
|---|---|
| Molecular Formula | C20H38O11 |
| Molecular Weight (g/mol) | 454.513 |
| MDL Number | MFCD00081763 |
| InChI Key | YLGXILFCIXHCMC-JHGZEJCSSA-N |
| PubChem CID | 51063134 |
| IUPAC Name | (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| SMILES | COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AA3671822
|
Thermo Scientific Chemicals
03671822 |
100 g |
Each for $73.99
|
|
|||||
|
AA3671836
|
Thermo Scientific Chemicals
03671836 |
500 g |
Each for $241.99
|
|
|||||
|
AA36718A4
|
Thermo Scientific Chemicals
036718A4 |
2.5 kg |
Each for $950.56
|
|
|||||
Description
Methyl cellulose is used as a thickener and an emulsifier in various food and cosmetic products; as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. In paper and textile industries, it is used as a sizing, thereby protecting the fibers from water or oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9004-67-5 | |
| 454.513 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| C20H38O11 | |
| MFCD00081763 | |
| 51063134 | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Specifications
| 9004-67-5 | |
| MFCD00081763 | |
| Soluble in cold water. | |
| COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC | |
| 454.513 | |
| Powder |
| C20H38O11 | |
| 14,6040 | |
| YLGXILFCIXHCMC-JHGZEJCSSA-N | |
| (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane | |
| 51063134 | |
| Methyl cellulose, Viscosity: 4000cPs |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only